missing translation for 'onlineSavingsMsg'
Learn More

(1R,2S)-(+)-10,2-Camphorsultam, 99%

Catalog No. p-7021694
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g
Catalog No. Quantity
AAA1413303 1 g
AAA1413306 5 g
2 options

Catalog No. AAA1413303

Supplier: Thermo Scientific Chemicals A1413303

Only null left

CAS: 108448-77-7 | C10H17NO2S | 215.31 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 108448-77-7
Molecular Formula C10H17NO2S
Molecular Weight (g/mol) 215.31
MDL Number MFCD00151762,MFCD00066271,MFCD00151762,MFCD00151762
InChI Key DPJYJNYYDJOJNO-UHFFFAOYNA-N
Synonym 6s,7as-8,8-dimethylhexahydro-3h-3a,6-methanobenzo c isothiazole 2,2-dioxide
PubChem CID 132274328
IUPAC Name 10,10-dimethyl-3λ⁶-thia-4-azatricyclo[5.2.1.0¹,⁵]decane-3,3-dione
SMILES CC1(C)C2CCC11CS(=O)(=O)NC1C2

Specifications

Melting Point 179°C to 186°C
Quantity 1 g
Beilstein 4675755
Optical Rotation +33° (c=1 in Ethanol)
Formula Weight 215.32
Percent Purity 99%
Chemical Name or Material (1R,2S)-(+)-10,2-Camphorsultam
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only