missing translation for 'onlineSavingsMsg'
Learn More

(1R,2S)-(+)-10,2-Camphorsultam, 99%

Catalog No. AAA1413303
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1413303 1 g
AAA1413306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1413303 Supplier Thermo Scientific Chemicals Supplier No. A1413303
Only null left

CAS: 108448-77-7 | C10H17NO2S | 215.31 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 108448-77-7
Molecular Formula C10H17NO2S
Molecular Weight (g/mol) 215.31
MDL Number MFCD00151762,MFCD00066271,MFCD00151762,MFCD00151762
InChI Key DPJYJNYYDJOJNO-UHFFFAOYNA-N
Synonym 6s,7as-8,8-dimethylhexahydro-3h-3a,6-methanobenzo c isothiazole 2,2-dioxide
PubChem CID 132274328
IUPAC Name 10,10-dimethyl-3λ⁶-thia-4-azatricyclo[5.2.1.0¹,⁵]decane-3,3-dione
SMILES CC1(C)C2CCC11CS(=O)(=O)NC1C2

Specifications

Melting Point 179°C to 186°C
Quantity 1 g
Beilstein 4675755
Optical Rotation +33° (c=1 in Ethanol)
Formula Weight 215.32
Percent Purity 99%
Chemical Name or Material (1R,2S)-(+)-10,2-Camphorsultam
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only