missing translation for 'onlineSavingsMsg'
Learn More

(1R,2R)-(-)-N,N'-Dimethylcyclohexane-1,2-diamine 94.0+%, TCI America™

Catalog No. D24591G
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D24591G 1 g
D2459100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D24591G Supplier TCI America Supplier No. D24591G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 68737-65-5
Molecular Formula C8H20N2
Molecular Weight (g/mol) 144.26
MDL Number MFCD00671527,MFCD03001702
InChI Key JRHPOFJADXHYBR-HTQZYQBOSA-P
Synonym 1r,2r-n,n'-dimethyl-1,2-cyclohexanediamine, 1r,2r-n1,n2-dimethylcyclohexane-1,2-diamine, 1r,2r---n,n'-dimethylcyclohexane-1,2-diamine, trans-n1,n2-dimethylcyclohexane-1,2-diamine, trans-1r,2r-n,n'-bismethyl-1,2-cyclohexanediamine, trans-n,n'-dimethylcyclohexane-1,2-diamine, trans-n,n'-dimethyl-1,2-cyclohexanediamine, trans-1,2-bis methylamino cyclohexane, trans-1r,2r-n,n'-bismethyl-1,2-cyclohexane diamine, 1r,2r---1,2-bis methylamino cyclohexane
PubChem CID 2733821
IUPAC Name (1R,2R)-N1,N2-dimethylcyclohexane-1,2-bis(aminium)
SMILES C[NH2+][C@@H]1CCCC[C@H]1[NH2+]C

Specifications

Melting Point 45°C
Quantity 1 g
UN Number 3259
Formula Weight 142.25
Percent Purity ≥94.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (1R,2R)-(-)-N,N′-Dimethylcyclohexane-1,2-diamine
TSCA No
Recommended Storage Refrigerator