missing translation for 'onlineSavingsMsg'
Learn More

(1R,2R)-N,N'-Dimethyl-N,N'-bis(3,3-dimethylbutyl)cyclohexane-1,2-diamine 97.0+%, TCI America™

Catalog No. d38081g
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D38081G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D38081G Supplier TCI America Supplier No. D38081G

Chemical Identifiers

CAS 644958-86-1
Molecular Formula C20H42N2
Molecular Weight (g/mol) 310.57
InChI Key VGCWVKVNKNXOGZ-QZTJIDSGSA-N
PubChem CID 44629980
IUPAC Name (1R,2R)-1-N,2-N-bis(3,3-dimethylbutyl)-1-N,2-N-dimethylcyclohexane-1,2-diamine
SMILES CC(C)(C)CCN(C)C1CCCCC1N(C)CCC(C)(C)C

Specifications

Color Yellow
Boiling Point 150°C
Quantity 1 g
Formula Weight 310.57
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material (1R,2R)-N,N′-Dimethyl-N,N′-bis(3,3-dimethylbutyl)cyclohexane-1,2-diamine