missing translation for 'onlineSavingsMsg'
Learn More

(1R,2R)-N,N'-Dimethyl-1,2-cyclohexanediamine, 95%

Catalog No. p-4112251
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC337830010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC337830010 Supplier Thermo Scientific Chemicals Supplier No. 337830010

CAS: 68737-65-5 | C8H20N2 | 144.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 68737-65-5
Molecular Formula C8H20N2
Molecular Weight (g/mol) 144.26
MDL Number MFCD00671527,MFCD03001702
InChI Key JRHPOFJADXHYBR-HTQZYQBOSA-P
Synonym 1r,2r-n,n'-dimethyl-1,2-cyclohexanediamine, 1r,2r-n1,n2-dimethylcyclohexane-1,2-diamine, 1r,2r---n,n'-dimethylcyclohexane-1,2-diamine, trans-n1,n2-dimethylcyclohexane-1,2-diamine, trans-1r,2r-n,n'-bismethyl-1,2-cyclohexanediamine, trans-n,n'-dimethylcyclohexane-1,2-diamine, trans-n,n'-dimethyl-1,2-cyclohexanediamine, trans-1,2-bis methylamino cyclohexane, trans-1r,2r-n,n'-bismethyl-1,2-cyclohexane diamine, 1r,2r---1,2-bis methylamino cyclohexane
PubChem CID 2733821
IUPAC Name (1R,2R)-1-N,2-N-dimethylcyclohexane-1,2-diamine
SMILES C[NH2+][C@@H]1CCCC[C@H]1[NH2+]C

Specifications

CAS Min % 94.0
CAS Max % 100.0
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula C6H10(NHCH3)2
Quantity 1 g
Specific Rotation − 143.00
Formula Weight 142.24
Percent Purity 95%
Physical Form Low Melting Solid
Chemical Name or Material (1R,2R)-N,N'-Dimethyl-1,2-cyclohexanediamine
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only