missing translation for 'onlineSavingsMsg'
Learn More

(1R,2R,5R)-(+)-2-Hydroxy-3-pinanone 98.0+%, TCI America™

Catalog No. H08625G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H08625G 5 g
H08621G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H08625G Supplier TCI America Supplier No. H08625G

Chemical Identifiers

CAS 24047-72-1
Molecular Formula C10H16O2
Molecular Weight (g/mol) 168.236
MDL Number MFCD00191522
InChI Key VZRRCQOUNSHSGB-BRDIYROLSA-N
Synonym (1R,2R,5R)-(+)-2-Hydroxy-2,6,6-trimethylbicyclo[3.1.1]heptan-3-one
PubChem CID 6553882
IUPAC Name (1R,4R,5R)-4-hydroxy-4,6,6-trimethylbicyclo[3.1.1]heptan-3-one
SMILES CC1(C2CC1C(C(=O)C2)(C)O)C

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 168.24
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material (1R,2R,5R)-(+)-2-Hydroxy-3-pinanone