missing translation for 'onlineSavingsMsg'
Learn More

(1R,2R,3S,5R)-(-)-2,3-Pinanediol, 99%, Thermo Scientific™

Catalog No. p-7037770
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5658003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
Catalog No. AAH5658003 missing translation for 'mfr' Thermo Scientific Chemicals missing translation for 'supplierNo' H5658003

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 22422-34-0
Molecular Formula C10H18O2
Molecular Weight (g/mol) 170.25
MDL Number MFCD09955216
InChI Key MOILFCKRQFQVFS-UHFFFAOYNA-N
Synonym 1r,2r,3s,5r---2,3-pinanediol, 1r,2r,3s,5r---pinanediol, 1r,2r,3s,5r-2,6,6-trimethylbicyclo 3.1.1 heptane-2,3-diol, --2,3-pinanediol, unii-r58l0w3a75, --cis-pinane-cis-2,3-diol, pinanediol, --2-hydroxyisopinocampheol, bicyclo 3.1.1 heptane-2,3-diol, 2,6,6-trimethyl-, 1r,2r,3s,5r
PubChem CID 6553875
IUPAC Name (1R,3S,4R,5R)-4,6,6-trimethylbicyclo[3.1.1]heptane-3,4-diol
SMILES CC1(C)C2CC1C(C)(O)C(O)C2

Specifications

Melting Point 57°C to 59°C
Boiling Point 101°C to 102°C (1 mmHg)
Flash Point 110°C (230°F)
Quantity 1 g
Beilstein 2039144
Optical Rotation −8.6° (c=6.5 in Toluene)
Formula Weight 170.25
Percent Purity 99%
Chemical Name or Material (1R,2R,3S,5R)-(-)-2,3-Pinanediol
missing translation for 'healthHazard1' H315-H319-H335
missing translation for 'healthHazard3' P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures