missing translation for 'onlineSavingsMsg'
Learn More

(1R,2R)-2-(Naphthalene-2,3-dicarboximido)cyclohexanecarboxylic Acid 98.0+%, TCI America™

Catalog No. n0713100mg
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N0713100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N0713100MG Supplier TCI America Supplier No. N0713100MG

Chemical Identifiers

CAS 642995-15-1
Molecular Formula C19H17NO4
Molecular Weight (g/mol) 323.348
InChI Key RPVXICJCKYVHHZ-CZUORRHYSA-N
Synonym N-[(1R,2R)-2-Carboxycyclohexyl]naphthalene-2,3-dicarboximide
PubChem CID 44629756
IUPAC Name (1R,2R)-2-(1,3-dioxobenzo[f]isoindol-2-yl)cyclohexane-1-carboxylic acid
SMILES C1CCC(C(C1)C(=O)O)N2C(=O)C3=CC4=CC=CC=C4C=C3C2=O

Specifications

Color White
Quantity 100 mg
Formula Weight 323.35
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material (1R,2R)-2-(Naphthalene-2,3-dicarboximido)cyclohexanecarboxylic Acid