missing translation for 'onlineSavingsMsg'
Learn More

(1R,2R)-2-Aminocyclopentanol Hydrochloride 98.0+%, TCI America™

Catalog No. A29905G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A29905G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A29905G Supplier TCI America Supplier No. A29905G
Only null left

Chemical Identifiers

CAS 68327-11-7
Molecular Formula C5H12ClNO
Molecular Weight (g/mol) 137.607
MDL Number MFCD09834692
InChI Key ZFSXKSSWYSZPGQ-TYSVMGFPSA-N
Synonym (1R,2R)-2-Amino-1-hydroxycyclopentane Hydrochloride
PubChem CID 12886906
IUPAC Name (1R,2R)-2-aminocyclopentan-1-ol;hydrochloride
SMILES C1CC(C(C1)O)N.Cl

Specifications

Melting Point 180°C
Quantity 5 g
Formula Weight 137.61
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material (1R,2R)-2-Aminocyclopentanol Hydrochloride
TSCA No
Recommended Storage Refrigerator