missing translation for 'onlineSavingsMsg'
Learn More

(1R,2R)-2-Aminocyclohexanecarboxylic Acid 98.0+%, TCI America™

Catalog No. A1688100MG
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A1688100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A1688100MG Supplier TCI America Supplier No. A1688100MG

missing translation for 'validMainframeMsg'

Unnatural amino acid derivative used in amino acid and peptide synthesis.

Chemical Identifiers

CAS 26685-83-6
Molecular Formula C7H13NO2
Molecular Weight (g/mol) 143.186
InChI Key USQHEVWOPJDAAX-PHDIDXHHSA-N
Synonym 1r,2r-2-aminocyclohexanecarboxylic acid, trans-2-amino-1-cyclohexanecarboxylic acid, trans-2-aminocyclohexanecarboxylic acid, 1r,2r-2-aminocyclohexane-1-carboxylic acid, trans-2-amino-cyclohexanecarboxylic acid, cyclohexanecarboxylic acid, 2-amino-, trans, cyclohexanecarboxylic acid, 2-amino-, 1r,2r, cyclohexanecarboxylic acid, 2-amino-, 1r,2r-rel, 2-nchca, 2-aminocyclohexanecarboxylic acid #
PubChem CID 2724640
IUPAC Name (1R,2R)-2-aminocyclohexane-1-carboxylic acid
SMILES C1CCC(C(C1)C(=O)O)N

Specifications

Color White-Yellow
Quantity 100 mg
Formula Weight 143.19
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material (1R,2R)-2-Aminocyclohexanecarboxylic Acid