missing translation for 'onlineSavingsMsg'
Learn More

(1R,2R)-(+)-1,2-Diphenylethylenediamine 98.0+%, TCI America™

Catalog No. D21761G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D21761G 1 g
D21765G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D21761G Supplier TCI America Supplier No. D21761G

Chemical Identifiers

CAS 35132-20-8
Molecular Formula C14H18N2
Molecular Weight (g/mol) 214.31
MDL Number MFCD00082769
InChI Key PONXTPCRRASWKW-ZIAGYGMSSA-P
Synonym 1r,2r-+-1,2-diphenylethylenediamine, 1r,2r-1,2-diphenylethane-1,2-diamine, 1r,2r-rel-1,2-diphenylethane-1,2-diamine, 1r,2r-1,2-diphenylethylenediamine, 1r,2r-+-1,2-diamino-1,2-diphenylethane, 1r,2r-1,2-diphenyl-1,2-ethanediamine, 1r,2r-+-1,2-diphenyl-1,2-ethanediamine, +-1,2-diphenylethylenediamine, +-stilbenediamine, r,r-dpen
PubChem CID 2724998
IUPAC Name (1R,2R)-1,2-diphenylethane-1,2-bis(aminium)
SMILES [NH3+][C@@H]([C@H]([NH3+])C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 83°C
Color White-Yellow
Quantity 1 g
Formula Weight 212.30
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material (1R,2R)-(+)-1,2-Diphenylethylenediamine