missing translation for 'onlineSavingsMsg'
Learn More

(1R,2R)-(+)-1,2-Diphenyl-1,2-ethanediamine, 98+%

Catalog No. p-121944
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC294680010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC294680010 Supplier Thermo Scientific Chemicals Supplier No. 294680010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 35132-20-8 | C14H18N2 | 214.31 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 35132-20-8
Molecular Formula C14H18N2
Molecular Weight (g/mol) 214.31
MDL Number MFCD00082769
InChI Key PONXTPCRRASWKW-ZIAGYGMSSA-P
Synonym 1r,2r-+-1,2-diphenylethylenediamine, 1r,2r-1,2-diphenylethane-1,2-diamine, 1r,2r-rel-1,2-diphenylethane-1,2-diamine, 1r,2r-1,2-diphenylethylenediamine, 1r,2r-+-1,2-diamino-1,2-diphenylethane, 1r,2r-1,2-diphenyl-1,2-ethanediamine, 1r,2r-+-1,2-diphenyl-1,2-ethanediamine, +-1,2-diphenylethylenediamine, +-stilbenediamine, r,r-dpen
PubChem CID 2724998
IUPAC Name (1R,2R)-1,2-diphenylethane-1,2-diamine
SMILES [NH3+][C@@H]([C@H]([NH3+])C1=CC=CC=C1)C1=CC=CC=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 81.0°C to 84.0°C
Color White to Yellow
Enantiomeric Excess 98.5% min.
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula (C6H5CH(NH2)-)2
Quantity 1 g
Specific Rotation Condition + 104.00 (25.00°C c=1.1,MeOH)
Specific Rotation + 104.00
Formula Weight 212.29
Percent Purity 99%,99% ee
Physical Form Crystalline Powder
Chemical Name or Material (1R,2R)-(+)-1,2-Diphenyl-1,2-ethanediamine, 99% (99% e.e.)
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
TSCA TSCA

RUO – Research Use Only