missing translation for 'onlineSavingsMsg'
Learn More

(1R,2R)-1,2-Cyclohexanedicarboxylic Acid 98.0+%, TCI America™

Catalog No. p-7125209
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C19531G 1 g
C19535G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C19531G Supplier TCI America Supplier No. C19531G
Only null left

Chemical Identifiers

CAS 46022-05-3
Molecular Formula C8H12O4
Molecular Weight (g/mol) 172.18
InChI Key QSAWQNUELGIYBC-PHDIDXHHSA-N
Synonym 1r,2r-cyclohexane-1,2-dicarboxylic acid, trans-1,2-cyclohexanedicarboxylic acid, trans-cyclohexane-1,2-dicarboxylic acid, 1r,2r---1,2-cyclohexanedicarboxylic acid, 1r,2r-1,2-cyclohexanedicarboxylic acid, 1,2-cyclohexanedicarboxylic acid, 1r,2r-rel, 1,2-cyclohexanedicarboxylic acid, 1r,2r, 1r.2r-cyclohexane-1,2-dicarboxylicacid, 1,2-cyclohexanedicarboxylic acid, trans, pubchem19603
PubChem CID 720895
IUPAC Name (1R,2R)-cyclohexane-1,2-dicarboxylic acid
SMILES C1CCC(C(C1)C(=O)O)C(=O)O

Specifications

Melting Point 184°C
Color White
Quantity 1 g
Formula Weight 172.18
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material (1R,2R)-1,2-Cyclohexanedicarboxylic Acid
EINECSNumber (3)-2678
TSCA No