missing translation for 'onlineSavingsMsg'
Learn More

(1R,2R)-(-)-1,2-Cyclohexanediamine 98.0+%, TCI America™

Catalog No. p-7124987
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C14475G 5 g
C144725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C14475G Supplier TCI America Supplier No. C14475G
Only null left

Chemical Identifiers

CAS 20439-47-8
Molecular Formula C6H14N2
Molecular Weight (g/mol) 114.19
MDL Number MFCD00062985,MFCD00063747
InChI Key SSJXIUAHEKJCMH-UHFFFAOYNA-N
Synonym 1r,2r---1,2-diaminocyclohexane, 1r,2r-cyclohexane-1,2-diamine, trans-1,2-diaminocyclohexane, trans-1,2-cyclohexanediamine, trans-cyclohexane-1,2-diamine, 1r,2r-1,2-diaminocyclohexane, 1r,2r---1,2-cyclohexanediamine, 1r,2r-diaminocyclohexane, r-dach, 1r,2r-1,2-cyclohexanediamine
PubChem CID 43806
IUPAC Name cyclohexane-1,2-diamine
SMILES NC1CCCCC1N

Specifications

Melting Point 43°C
Color White-Yellow
Boiling Point 78°C
Quantity 5 g
UN Number 3259
Formula Weight 114.19
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Lumps
Chemical Name or Material (1R,2R)-(-)-1,2-Cyclohexanediamine
EINECSNumber (3)-4230
TSCA No