missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(1R,2R)-(-)-1,2-Cyclohexanediamine 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 43°C |
| Color | White-Yellow |
| Boiling Point | 78°C |
| Quantity | 25 g |
| UN Number | 3259 |
| Formula Weight | 114.19 |
| Percent Purity | ≥98.0% (GC,T) |
| Physical Form | Crystalline Lumps |
| Chemical Name or Material | (1R,2R)-(-)-1,2-Cyclohexanediamine |
Safety and Handling
| EINECSNumber | (3)-4230 |
| TSCA | No |