Learn More
Spectracer PERCHLORIC ACID-DIOXANE,1000ML
Catalog No. NC1455955
Encompass
Perchloric Acid in DioxaneConcentration: Tenth-Normal (0.1 N)Volume (ml): 1000ml Glass Bottle(s)Shelf Life: 12 Months.
Catalog No. NC1455955 Supplier Spectracer Supplier No. USPVS 114
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Perchloric Acid in DioxaneConcentration: Tenth-Normal (0.1 N)Volume (ml): 1000ml Glass Bottle(s)Shelf Life: 12 Months.