missing translation for 'onlineSavingsMsg'
Learn More

1H-Pyrazolo[3,4-b]pyridine, 97%

Catalog No. aah6171003
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6171003 1 g
AAH61710MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6171003 Supplier Thermo Scientific Chemicals Supplier No. H6171003
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 271-73-8 | C6H5N3 | 119.127 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 271-73-8
Molecular Formula C6H5N3
Molecular Weight (g/mol) 119.127
MDL Number MFCD05663981
InChI Key GVLRTOYGRNLSDW-UHFFFAOYSA-N
Synonym 1h-pyrazolo 3,4-b pyridine, 2h-pyrazolo 3,4-b pyridine, 7-azaindazole, pyrazolo 3,4-b pyridine, 7-aza-1h-indazole, pyrazolo 5,4-b pyridine, 7-aza indazole, 7-azabenzpyrazole, acmc-1cs3b, 2h pyrazolo 3,4 b pyridin
PubChem CID 2755850
IUPAC Name 1H-pyrazolo[3,4-b]pyridine
SMILES C1=CC2=C(NN=C2)N=C1

Specifications

Quantity 1 g
Formula Weight 119.12
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 1H-Pyrazolo[3,4-b]pyridine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only