missing translation for 'onlineSavingsMsg'
Learn More

1H-Pyrazole-4-boronic acid pinacol ester, 98%

Catalog No. p-7047710
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1965403 1 g
AAL1965406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1965403 Supplier Thermo Scientific Chemicals Supplier No. L1965403
Only null left

CAS: 269410-08-4 | C9H15BN2O2 | 194.04 g/mol

1H-Pyrazole-4-boronic acid pinacol ester is a useful reagent for Suzuki-Miyaura cross-couplings as well as Ruthenium-catalyzed asymmetric hydrogenation. It is also used as reagent for preparing VEGF, Aurora, RHO (ROCK), Janus Kinase 2, c-MET, ALK, S-nitrsoflutathione reductase, CDC7, Acetyl-CoA carboxylase inhibitors as well as other biologically active compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1H-Pyrazole-4-boronic acid pinacol ester is a useful reagent for Suzuki-Miyaura cross-couplings as well as Ruthenium-catalyzed asymmetric hydrogenation. It is also used as reagent for preparing VEGF, Aurora, RHO (ROCK), Janus Kinase 2, c-MET, ALK, S-nitrsoflutathione reductase, CDC7, Acetyl-CoA carboxylase inhibitors as well as other biologically active compounds.

Solubility
Insoluble in water.

Notes
Keep the container tightly sealed. Store in a cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 269410-08-4
Molecular Formula C9H15BN2O2
Molecular Weight (g/mol) 194.04
MDL Number MFCD03453063
InChI Key TVOJIBGZFYMWDT-UHFFFAOYSA-N
Synonym 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole, pyrazole-4-boronic acid pinacol ester, 4-pyrazoleboronic acid pinacol ester, 1h-pyrazole-4-boronic acid pinacol ester, 4-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole, 1h-pyrazole-4-boronic acid, pinacol ester, 4,4,5,5-tetramethyl-2-1h-pyrazol-4-yl-1,3,2-dioxaborolane, pyrazole-4-boronic acid, pinacol ester, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrazole, 2-4-pyrazolyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
PubChem CID 2774010
IUPAC Name 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole
SMILES CC1(C)OB(OC1(C)C)C1=CNN=C1

Specifications

Melting Point 145°C to 149°C
Quantity 1 g
Solubility Information Insoluble in water.
Formula Weight 194.04
Percent Purity 98%
Chemical Name or Material 1H-Pyrazole-4-boronic acid pinacol ester
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only