missing translation for 'onlineSavingsMsg'
Learn More

1H-Pyrazole-3-carboxylic acid, 97%

Catalog No. AAB2573203
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2573203 1 g
AAB2573206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2573203 Supplier Thermo Scientific Chemicals Supplier No. B2573203
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1621-91-6 | C4H4N2O2 | 112.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1621-91-6
Molecular Formula C4H4N2O2
Molecular Weight (g/mol) 112.09
MDL Number MFCD00077436
InChI Key KOPFEFZSAMLEHK-UHFFFAOYSA-N
Synonym 1h-pyrazole-3-carboxylic acid, pyrazole-3-carboxylic acid, 2h-pyrazole-3-carboxylic acid, 5-pyrazolecarboxylic acid, 3-pyrazolecarboxylic acid, 3-carboxy-1h-pyrazole, pyrazolic acid, pyrazolecarboxylic, 3-carboxypyrazole, 2h-pyrazole-3-carboxylicacid
PubChem CID 574310
IUPAC Name 1H-pyrazole-5-carboxylic acid
SMILES OC(=O)C1=CC=NN1

Specifications

Melting Point 211°C (decomposition)
Quantity 1 g
Formula Weight 112.09
Percent Purity 97%
Chemical Name or Material 1H-Pyrazole-3-carboxylic acid
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only