missing translation for 'onlineSavingsMsg'
Learn More

1H-Pyrazole-3,5-dicarboxylic acid monohydrate, 98%

Catalog No. p-7044489
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0673106 5 g
AAL0673114 25 g
AAL0673122 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL0673106 Supplier Thermo Scientific Chemicals Supplier No. L0673106

CAS: 303180-11-2 | C5H6N2O5 | 174.112 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 303180-11-2
Molecular Formula C5H6N2O5
Molecular Weight (g/mol) 174.112
MDL Number MFCD00149323
InChI Key GLINCONFUZIMCN-UHFFFAOYSA-N
Synonym 1h-pyrazole-3,5-dicarboxylic acid hydrate, 3,5-pyrazoledicarboxylic acid monohydrate, 1h-pyrazole-3,5-dicarboxylic acid monohydrate, pyrazole-3,5-dicarboxylic acid, hydrate, acmc-1aezk, 3,5-pyrazoledicarboxylic acid hydrate, 1h-pyrazole-3,5-dicarboxylicacidmonohydrate, 1h-pyrazole-3,5-dicarboxylic acid; hydrate, 1h-pyrazole-3,5-dicarboxylic acid hydrate 1:1
PubChem CID 2723723
IUPAC Name 1H-pyrazole-3,5-dicarboxylic acid;hydrate
SMILES C1=C(NN=C1C(=O)O)C(=O)O.O

Specifications

Melting Point ∽294°C (decomposition)
Quantity 5 g
Beilstein 147821
Formula Weight 174.11 (156.09 Anhydrous)
Percent Purity 98%
Chemical Name or Material 1H-Pyrazole-3,5-dicarboxylic acid monohydrate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 221-474-7
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only