missing translation for 'onlineSavingsMsg'
Learn More

1H-Pyrazol-4-amine 98.0+%, TCI America™

Catalog No. p25231g
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P25231G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P25231G Supplier TCI America Supplier No. P25231G
Only null left

Chemical Identifiers

CAS 28466-26-4
Molecular Formula C3H5N3
Molecular Weight (g/mol) 83.094
MDL Number MFCD01693729
InChI Key AXINVSXSGNSVLV-UHFFFAOYSA-N
Synonym 4-aminopyrazole, 4-amino-1h-pyrazole, 1h-pyrazol-4-ylamine, 9ci, pyrazole, 4-amino, 1h-pyrazole-4-amine, pyrazole-4-ylamine, 4-nh2-pyrazole, 1-h-pyrazol-4-amine, pubchem14988
PubChem CID 78035
IUPAC Name 1H-pyrazol-4-amine
SMILES C1=C(C=NN1)N

Specifications

Melting Point 84°C
Color Brown-Yellow
Quantity 1 g
Formula Weight 83.09
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1H-Pyrazol-4-amine