missing translation for 'onlineSavingsMsg'
Learn More

1H-Indene-3-carboxylic acid, 97%

Catalog No. p-7032753
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3434403 1 g
AAH3434406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3434403 Supplier Thermo Scientific Chemicals Supplier No. H3434403

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 5020-21-3 | C10H8O2 | 160.172 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5020-21-3
Molecular Formula C10H8O2
Molecular Weight (g/mol) 160.172
MDL Number MFCD00086207
InChI Key NZSCUDBGUBVDLO-UHFFFAOYSA-N
Synonym 1h-indene-3-carboxylic acid, indene-3-carboxylic acid, 3-indenecarboxylic acid
PubChem CID 84261
IUPAC Name 3H-indene-1-carboxylic acid
SMILES C1C=C(C2=CC=CC=C21)C(=O)O

Specifications

Melting Point 161°C to 165°C
Quantity 1 g
Formula Weight 160.17
Percent Purity 97%
Chemical Name or Material 1H-Indene-3-carboxylic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only