missing translation for 'onlineSavingsMsg'
Learn More

1H-Indazole-6-carboxaldehyde, 97%

Catalog No. AC431040010
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC431040010 1 g Glass bottle
AC431040050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC431040010 Supplier Thermo Scientific Chemicals Supplier No. 431040010

CAS: 669050-69-5 | C8H6N2O | 146.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 669050-69-5
Molecular Formula C8H6N2O
Molecular Weight (g/mol) 146.15
InChI Key JTWYTTXTJFDYAG-UHFFFAOYSA-N
Synonym 1h-indazole-6-carboxaldehyde, 6-formylindazole, 6-formyl-1h-indazole, 2h-indazole-6-carbaldehyde, indazole-6-carboxaldehyde, 1h-indazole-6-cabaldehyde, pubchem23878, acmc-1bc45, 2h-indazole-6-carboxaldehyde, 1-h-indazole-6-carboxaldehyde
PubChem CID 18538368
IUPAC Name 1H-indazole-6-carbaldehyde
SMILES C1=CC2=C(C=C1C=O)NN=C2

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 180.6°C to 180.8°C
Color Brown to White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 146.15
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 1H-Indazole-6-carboxaldehyde
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Take off contaminated clothing and wash before reuse.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water

RUO – Research Use Only