missing translation for 'onlineSavingsMsg'
Learn More

1H-indazole-3-carboxylic acid, Thermo Scientific™

Catalog No. MO07468DA
Change view
Click to view available options
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
MO07468DA 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MO07468DA Supplier Thermo Scientific Chemicals Supplier No. MO07468DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 4498-67-3
Assay Percent Range 97%
Molecular Formula C8H6N2O2
Quantity 1g
Synonym indazole-3-carboxylic acid, 3-carboxyindazole, 3-indazolecarboxylic acid, indazole-3-carboxylicacid, 1h-indazol-3-carbonic acid, 2h-indazole-3-carboxylic acid, 3-carboxy-1h-indazole, 1h-indazole-3-carbonic acid, 1-h-indazole-3-carboxylic acid, benzopyrazole-3-carboxylic acid
InChI Key BHXVYTQDWMQVBI-UHFFFAOYSA-N
SMILES C1=CC=C2C(=C1)C(=NN2)C(=O)O
IUPAC Name 1H-indazole-3-carboxylic acid
Molecular Weight (g/mol) 162.148
PubChem CID 78250
Formula Weight 162.15
Percent Purity 97%
Chemical Name or Material 1H-Indazole-3-carboxylic acid
Show More Show Less