missing translation for 'onlineSavingsMsg'
Learn More

1H-Indazole-3-carboxylic acid, 98%

Catalog No. p-7055423
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6400106 5 g
AAH6400114 25 g
AAH6400122 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH6400106 Supplier Thermo Scientific Chemicals Supplier No. H6400106

CAS: 4498-67-3 | C8H6N2O2 | 162.148 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4498-67-3
Molecular Formula C8H6N2O2
Molecular Weight (g/mol) 162.148
MDL Number MFCD00211066
InChI Key BHXVYTQDWMQVBI-UHFFFAOYSA-N
Synonym indazole-3-carboxylic acid, 3-carboxyindazole, 3-indazolecarboxylic acid, indazole-3-carboxylicacid, 1h-indazol-3-carbonic acid, 2h-indazole-3-carboxylic acid, 3-carboxy-1h-indazole, 1h-indazole-3-carbonic acid, 1-h-indazole-3-carboxylic acid, benzopyrazole-3-carboxylic acid
PubChem CID 78250
IUPAC Name 1H-indazole-3-carboxylic acid
SMILES C1=CC=C2C(=C1)C(=NN2)C(=O)O

Specifications

Melting Point 266°C to 270°C (literature)
Color Brown
Quantity 5 g
Beilstein 6354
Formula Weight 162.15
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 1H-Indazole-3-carboxylic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 224-794-5
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only