missing translation for 'onlineSavingsMsg'
Learn More

1H-Imidazole-4-carboxylic acid, 98%

Catalog No. AC397110010
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC397110010 1 g Glass bottle
AC397110050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC397110010 Supplier Thermo Scientific Chemicals Supplier No. 397110010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1072-84-0 | C4H4N2O2 | 112.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1072-84-0
Molecular Formula C4H4N2O2
Molecular Weight (g/mol) 112.09
MDL Number MFCD00082203
InChI Key NKWCGTOZTHZDHB-UHFFFAOYSA-N
Synonym 1h-imidazole-4-carboxylic acid, 4-imidazolecarboxylic acid, imidazole-4-carboxylic acid, 3h-imidazole-4-carboxylic acid, 4-carboxyimidazole, unii-ae82z8u4gj, imidazole-5-carboxylic acid, 4-imidazole carboxylic acid, ae82z8u4gj, 4 5-imidazolecarboxylic acid
PubChem CID 14080
IUPAC Name 1H-imidazole-5-carboxylic acid
SMILES OC(=O)C1=CN=CN1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 242°C to 244°C
Color Colorless
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 112.09
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1H-Imidazole-4-carboxylic acid
Show More Show Less
Hazard Statement GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 214-017-8

RUO – Research Use Only