missing translation for 'onlineSavingsMsg'
Learn More

1H-Imidazole-4-carbaldehyde, 97%, Thermo Scientific™

Catalog No. CC08804EB
Change view
Click to view available options
Quantity:
25 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC08804EB 25 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CC08804EB Supplier Thermo Scientific Chemicals Supplier No. CC08804EB

Chemical Identifiers

CAS 3034-50-2
Molecular Formula C4H4N2O
Molecular Weight (g/mol) 96.089
MDL Number MFCD00173726
InChI Key ZQEXIXXJFSQPNA-UHFFFAOYSA-N
Synonym 1h-imidazole-4-carbaldehyde, imidazole-4-carbaldehyde, 1h-imidazole-4-carboxaldehyde, imidazole-4-carboxaldehyde, 4-imidazolecarboxaldehyde, 4-formylimidazole, 4 5-imidazolecarboxaldehyde, 3h-imidazole-4-carbaldehyde, 4-formyl-1h-imidazole, imidazole-5-carbaldehyde
PubChem CID 76428
IUPAC Name 1H-imidazole-5-carbaldehyde
SMILES C1=C(NC=N1)C=O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 25 g
Formula Weight 96.09
Percent Purity 97%
Chemical Name or Material 1H-imidazole-4-carbaldehyde
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful in contact with skin.
May cause respiratory irritation.
Causes skin irritation.
Harmful if swallowed.
Harmful if inhaled.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.