missing translation for 'onlineSavingsMsg'
Learn More

1H-Imidazole-4,5-dicarboxylic Acid 98.0+%, TCI America™

Catalog No. I00035G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I00035G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I00035G Supplier TCI America Supplier No. I00035G

Chemical Identifiers

CAS 570-22-9
Molecular Formula C5H4N2O4
Molecular Weight (g/mol) 156.10
MDL Number MFCD00005200
InChI Key ZEVWQFWTGHFIDH-UHFFFAOYSA-N
Synonym 4,5-imidazoledicarboxylic acid, imidazole-4,5-dicarboxylic acid, glyoxalinedicarboxylic acid, 4,5-dicarboxyimidazole, 4,5-imidazoledicarboxylate, glycoxalinedicarboxylic acid, alpha-beta-imidazolecarboxylic acid, alpha,beta-imidazoledicarboxylic acid, 4.5-imidazoledicarboxylic acid, 4,5-imidazole dicarboxylic acid
PubChem CID 68442
IUPAC Name 1H-imidazole-4,5-dicarboxylic acid
SMILES OC(=O)C1=C(N=CN1)C(O)=O

Specifications

Melting Point 283°C
Color White-Yellow
Quantity 5 g
Formula Weight 156.10
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1H-Imidazole-4,5-dicarboxylic Acid
EINECSNumber (5)-6853
RTECSNumber NI4760000
TSCA Yes