missing translation for 'onlineSavingsMsg'
Learn More

1H-Imidazol-2-ylmethanol hydrochloride, 97%, Thermo Scientific™

Catalog No. p-2695277
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC08909DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CC08909DA Supplier Thermo Scientific Chemicals Supplier No. CC08909DA

Chemical Identifiers

CAS 116177-22-1
Molecular Formula C4H7ClN2O
Molecular Weight (g/mol) 134.563
MDL Number MFCD03659700
InChI Key FEORNCWZOSTSRO-UHFFFAOYSA-N
Synonym 1h-imidazol-2-ylmethanol hydrochloride, 1h-imidazol-2-yl methanol hydrochloride, imidazolylmethanol hydrochloride, hydroxymethylimidazole hydrochloride, hydroxymethyl imidazole hydrochloride, imidazol-2-ylmethan-1-ol, chloride
PubChem CID 2776278
IUPAC Name 1H-imidazol-2-ylmethanol;hydrochloride
SMILES C1=CN=C(N1)CO.Cl

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 134.57
Percent Purity 97%
Chemical Name or Material 1H-imidazol-2-ylmethanol hydrochloride
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.