missing translation for 'onlineSavingsMsg'
Learn More

1H-Benzotriazole-1-carboxaldehyde 95.0+%, TCI America™

Catalog No. B39205G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B39205G 5 g
B39201G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B39205G Supplier TCI America Supplier No. B39205G
Only null left

Chemical Identifiers

CAS 72773-04-7
Molecular Formula C7H5N3O
Molecular Weight (g/mol) 147.137
MDL Number MFCD00239420
InChI Key DVEIDGKSJOJJJU-UHFFFAOYSA-N
Synonym 1-Formyl-1H-benzotriazole
PubChem CID 576740
IUPAC Name benzotriazole-1-carbaldehyde
SMILES C1=CC=C2C(=C1)N=NN2C=O

Specifications

Melting Point 97°C
Color White
Quantity 5 g
Formula Weight 147.14
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1H-Benzotriazole-1-carboxaldehyde
TSCA No
Recommended Storage Refrigerator