missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1H-Benzotriazole-1-carboxaldehyde 95.0+%, TCI America™
Specifications
Specifications
| Melting Point | 97°C |
| Color | White |
| Quantity | 1 g |
| Formula Weight | 147.14 |
| Percent Purity | ≥95.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 1H-Benzotriazole-1-carboxaldehyde |
Safety and Handling
| TSCA | No |
| Recommended Storage | Refrigerator |