missing translation for 'onlineSavingsMsg'
Learn More

1H-Benzotriazol-1-yloxytripyrrolidinophosphonium Hexafluorophosphate 98.0+%, TCI America™

Catalog No. p-7119256
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B17745G 5 g
B177425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B17745G Supplier TCI America Supplier No. B17745G

Chemical Identifiers

CAS 128625-52-5
Molecular Formula C18H28F6N6OP2
Molecular Weight (g/mol) 520.401
MDL Number MFCD00077411
InChI Key VIAFLMPQBHAMLI-UHFFFAOYSA-N
Synonym pybop, 1h-benzo d 1,2,3 triazol-1-yl oxy tri pyrrolidin-1-yl phosphonium hexafluorophosphate v, benzotriazol-1-yloxy tripyrrolidinophosphonium hexafluorophosphate, benzotriazole-1-yl-oxytripyrrolidinophosphonium hexafluorophosphate, benzotriazol-1-yl-oxytripyrrolidinophosphonium hexafluorophosphate, 1h-benzotriazol-1-yloxytripyrrolidinophosphonium hexafluorophosphate, benzotriazol-1-yloxytris pyrrolidino phosphonium hexafluorophosphate, benzotriazole-1-yl-oxy-tris-pyrrolidino-phosphonium hexafluorophosphate, benzotriazol-1-yl-oxy-tripyrrolidinophosphonium hexafluorophosphate, benzotriazol-1-yl-oxytripyrrolidinphosphonium hexafluorophosphate
PubChem CID 2724699
IUPAC Name benzotriazol-1-yloxy(tripyrrolidin-1-yl)phosphanium;hexafluorophosphate
SMILES C1CCN(C1)[P+](N2CCCC2)(N3CCCC3)ON4C5=CC=CC=C5N=N4.F[P-](F)(F)(F)(F)F

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 520.40
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 1H-Benzotriazol-1-yloxytripyrrolidinophosphonium Hexafluorophosphate [Coupling Reagent for Peptide]
TSCA No
Recommended Storage Refrigerator