missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1H-Benzotriazol-1-yloxytri(1-pyrrolidinyl)phosphonium hexafluorophosphate, 98%
Description
1H-Benzotriazol-1-yloxytri(1-pyrrolidinyl) phosphonium hexafluorophosphate is a peptide coupling reagent utilized in solid phase peptide synthesis. It is also used as a substitute for the (benzotriazol-1-yloxy)tris(dimethylamino)phosphonium hexafluorophosphate (BOP) reagent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1H-Benzotriazol-1-yloxytri(1-pyrrolidinyl) phosphonium hexafluorophosphate is a peptide coupling reagent utilized in solid phase peptide synthesis. It is also used as a substitute for the (benzotriazol-1-yloxy)tris(dimethylamino)phosphonium hexafluorophosphate (BOP) reagent.
Solubility
Soluble in chloroform.
Notes
Store in a cool place. Incompatible with oxidizing agents and water.
1H-Benzotriazol-1-yloxytri(1-pyrrolidinyl) phosphonium hexafluorophosphate is a peptide coupling reagent utilized in solid phase peptide synthesis. It is also used as a substitute for the (benzotriazol-1-yloxy)tris(dimethylamino)phosphonium hexafluorophosphate (BOP) reagent.
Solubility
Soluble in chloroform.
Notes
Store in a cool place. Incompatible with oxidizing agents and water.
Specifications
Specifications
| Melting Point | ∼155°C (decomposition) |
| Quantity | 5 g |
| Beilstein | 3584612 |
| Sensitivity | Moisture sensitive |
| Solubility Information | Soluble in chloroform. |
| Formula Weight | 520.4 |
| Percent Purity | 98% |
| Chemical Name or Material | 1H-Benzotriazol-1-yloxytri(1-pyrrolidinyl)phosphonium hexafluorophosphate |
Safety and Handling
| Hazard Category | H315-H319-H335 |
| Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P305+P351+P338-P312-P332+P313-P362-P501c |
| EINECSNumber | 000-000-0 |
| TSCA | No |
| Recommended Storage | Keep cold |
RUO – Research Use Only