missing translation for 'onlineSavingsMsg'
Learn More

1H-Benzotriazol-1-yloxytri(1-pyrrolidinyl)phosphonium hexafluorophosphate, 98%

Catalog No. AAB2525106
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2525106 5 g
AAB2525114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2525106 Supplier Thermo Scientific Chemicals Supplier No. B2525106

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 128625-52-5 | C18H28F6N6OP2 | 520.401 g/mol

1H-Benzotriazol-1-yloxytri(1-pyrrolidinyl) phosphonium hexafluorophosphate is a peptide coupling reagent utilized in solid phase peptide synthesis. It is also used as a substitute for the (benzotriazol-1-yloxy)tris(dimethylamino)phosphonium hexafluorophosphate (BOP) reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1H-Benzotriazol-1-yloxytri(1-pyrrolidinyl) phosphonium hexafluorophosphate is a peptide coupling reagent utilized in solid phase peptide synthesis. It is also used as a substitute for the (benzotriazol-1-yloxy)tris(dimethylamino)phosphonium hexafluorophosphate (BOP) reagent.

Solubility
Soluble in chloroform.

Notes
Store in a cool place. Incompatible with oxidizing agents and water.

Chemical Identifiers

CAS 128625-52-5
Molecular Formula C18H28F6N6OP2
Molecular Weight (g/mol) 520.401
MDL Number MFCD00077411
InChI Key VIAFLMPQBHAMLI-UHFFFAOYSA-N
Synonym pybop, 1h-benzo d 1,2,3 triazol-1-yl oxy tri pyrrolidin-1-yl phosphonium hexafluorophosphate v, benzotriazol-1-yloxy tripyrrolidinophosphonium hexafluorophosphate, benzotriazole-1-yl-oxytripyrrolidinophosphonium hexafluorophosphate, benzotriazol-1-yl-oxytripyrrolidinophosphonium hexafluorophosphate, 1h-benzotriazol-1-yloxytripyrrolidinophosphonium hexafluorophosphate, benzotriazol-1-yloxytris pyrrolidino phosphonium hexafluorophosphate, benzotriazole-1-yl-oxy-tris-pyrrolidino-phosphonium hexafluorophosphate, benzotriazol-1-yl-oxy-tripyrrolidinophosphonium hexafluorophosphate, benzotriazol-1-yl-oxytripyrrolidinphosphonium hexafluorophosphate
PubChem CID 2724699
IUPAC Name benzotriazol-1-yloxy(tripyrrolidin-1-yl)phosphanium;hexafluorophosphate
SMILES C1CCN(C1)[P+](N2CCCC2)(N3CCCC3)ON4C5=CC=CC=C5N=N4.F[P-](F)(F)(F)(F)F

Specifications

Melting Point ∼155°C (decomposition)
Quantity 5 g
Beilstein 3584612
Sensitivity Moisture sensitive
Solubility Information Soluble in chloroform.
Formula Weight 520.4
Percent Purity 98%
Chemical Name or Material 1H-Benzotriazol-1-yloxytri(1-pyrrolidinyl)phosphonium hexafluorophosphate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only