missing translation for 'onlineSavingsMsg'
Learn More

1H,1H-Tridecafluoroheptyl Iodide 95.0+%, TCI America™

Catalog No. T24825G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T24825G 5 g
T24821G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T24825G Supplier TCI America Supplier No. T24825G
Only null left

Chemical Identifiers

CAS 212563-43-4
Molecular Formula C7H2F13I
Molecular Weight (g/mol) 459.977
MDL Number MFCD00233026
InChI Key GBDROMNKGSCMDH-UHFFFAOYSA-N
Synonym 1H,1H-Perfluoro-1-iodoheptane, 1H,1H-Tridecafluoro-1-iodoheptane, 1H,1H-Perfluoroheptyl Iodide
PubChem CID 2782379
IUPAC Name 1,1,1,2,2,3,3,4,4,5,5,6,6-tridecafluoro-7-iodoheptane
SMILES C(C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)I

Specifications

Color Yellow
Quantity 5 g
Formula Weight 459.98
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 1H,1H-Tridecafluoroheptyl Iodide