missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1H,1H-Tridecafluoro-1-heptanol 95.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Boiling Point | 95°C |
| Quantity | 5 g |
| Formula Weight | 350.08 |
| Percent Purity | ≥95.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | 1H,1H-Tridecafluoro-1-heptanol |
Safety and Handling
| TSCA | No |