missing translation for 'onlineSavingsMsg'
Learn More

1H,1H-Perfluoro-1-dodecanol, tech. 90%

Catalog No. p-7046855
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1659006 5 g
AAL1659014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1659006 Supplier Thermo Scientific Chemicals Supplier No. L1659006

CAS: 423-65-4 | C12H3F23O | 600.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 423-65-4
Molecular Formula C12H3F23O
Molecular Weight (g/mol) 600.12
MDL Number MFCD00153235
InChI Key SHTZQFTXUMCALC-UHFFFAOYSA-N
Synonym 1h,1h-perfluoro-1-dodecanol, 1h,1h-perfluorododecan-1-ol, 1h,1h-perfluorododecanol, tricosafluoroundecylmethanol, 1h,1h-tricosafluoro-1-dodecanol, 1h,1h-perfluoro-1-lauryl alcohol
PubChem CID 2760321
IUPAC Name 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,12-tricosafluorododecan-1-ol
SMILES OCC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F

Specifications

Melting Point ∼110°C
Boiling Point 224°C (740 mmHg)
Quantity 5 g
Beilstein 1810966
Formula Weight 600.12
Percent Purity 90%
Grade Technical
Chemical Name or Material 1H,1H-Perfluoro-1-dodecanol
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only