missing translation for 'onlineSavingsMsg'
Learn More

1H,1H-Perfluoro-1-decanol, 98%

Catalog No. p-7046849
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1658006 5 g
AAL1658014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1658006 Supplier Thermo Scientific Chemicals Supplier No. L1658006
Only null left

CAS: 307-37-9 | C10H3F19O | 500.103 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 307-37-9
Molecular Formula C10H3F19O
Molecular Weight (g/mol) 500.103
MDL Number MFCD00153234
InChI Key NIRPXSQCRWXHNZ-UHFFFAOYSA-N
Synonym 1h,1h-perfluoro-1-decanol, 1h,1h-nonadecafluoro-1-decanol, 1h,1h-perfluorodecan-1-ol, nonadecafluorononyl methanol, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-nonadecakis fluoranyl decan-1-ol
PubChem CID 2733275
IUPAC Name 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-nonadecafluorodecan-1-ol
SMILES C(C(C(C(C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)O

Specifications

Melting Point 83°C to 90°C
Boiling Point 95°C (20 mmHg)
Linear Formula CF3(CF2)8CH2OH
Quantity 5 g
Beilstein 1810012
Formula Weight 500.1
Percent Purity 98%
Chemical Name or Material 1H,1H-Perfluoro-1-decanol
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only