missing translation for 'onlineSavingsMsg'
Learn More

1H,1H-Pentadecafluoro-n-octyl Acrylate (stabilized with MEHQ) 95.0+%, TCI America™

Supplier:  TCI America P175425G

 View more versions of this product

Catalog No. P175425G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

1H,1H-Pentadecafluoro-n-octyl Acrylate (stabilized with MEHQ)
Yellow
C11H5F15O2
25 g
YSQGYEYXKXGAQA-UHFFFAOYSA-N
2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecafluorooctyl prop-2-enoate
67551
≥95.0% (GC)
307-98-2
65°C
MFCD00039248
1h,1h-pentadecafluorooctyl acrylate, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecafluorooctyl acrylate, 1h,1h-perfluorooctyl acrylate, acrylic acid 1h,1h-pentadecafluoro-n-octyl ester, 2-propenoic acid, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecafluorooctyl ester, 1h,1h-perfluoro-n-octyl acrylate, 1h,1h-pentadecafluoro-n-octyl acrylate, 1,1-dihydroperfluorooctyl acrylate, 1,1-dyhydroperfluorooctyl acrylate, acrylic acid 1h,1h-perfluorooctyl ester
C=CC(=O)OCC(C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F
454.135
454.14
Liquid

Chemical Identifiers

307-98-2
454.135
YSQGYEYXKXGAQA-UHFFFAOYSA-N
67551
C=CC(=O)OCC(C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F
C11H5F15O2
MFCD00039248
1h,1h-pentadecafluorooctyl acrylate, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecafluorooctyl acrylate, 1h,1h-perfluorooctyl acrylate, acrylic acid 1h,1h-pentadecafluoro-n-octyl ester, 2-propenoic acid, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecafluorooctyl ester, 1h,1h-perfluoro-n-octyl acrylate, 1h,1h-pentadecafluoro-n-octyl acrylate, 1,1-dihydroperfluorooctyl acrylate, 1,1-dyhydroperfluorooctyl acrylate, acrylic acid 1h,1h-perfluorooctyl ester
2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecafluorooctyl prop-2-enoate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions