missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1H,1H-Pentadecafluoro-1-octanol 98.0+%, TCI America™
$92.27 - $325.46
Chemical Identifiers
| CAS | 307-30-2 |
|---|---|
| Molecular Formula | C8H3F15O |
| Molecular Weight (g/mol) | 400.087 |
| MDL Number | MFCD00004675 |
| InChI Key | PJDOLCGOTSNFJM-UHFFFAOYSA-N |
| Synonym | 1h,1h-pentadecafluoro-1-octanol, 1h,1h-perfluoro-1-octanol, 1h,1h-perfluorooctanol, pentadecafluorooctyl alcohol, unii-9x5w7fq62r, 1-octanol, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecafluoro, pentadecafluoro-1-octanol, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecafluoro-1-octanol, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecafluorooctyl alcohol, perfluorooctanol |
| PubChem CID | 67543 |
| IUPAC Name | 2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecafluorooctan-1-ol |
| SMILES | C(C(C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)O |
Chemical Identifiers
| 307-30-2 | |
| 400.087 | |
| PJDOLCGOTSNFJM-UHFFFAOYSA-N | |
| 67543 | |
| C(C(C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)O |
| C8H3F15O | |
| MFCD00004675 | |
| 1h,1h-pentadecafluoro-1-octanol, 1h,1h-perfluoro-1-octanol, 1h,1h-perfluorooctanol, pentadecafluorooctyl alcohol, unii-9x5w7fq62r, 1-octanol, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecafluoro, pentadecafluoro-1-octanol, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecafluoro-1-octanol, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecafluorooctyl alcohol, perfluorooctanol | |
| 2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecafluorooctan-1-ol |
Specifications
| 307-30-2 | |
| 164°C | |
| MFCD00004675 | |
| 1h,1h-pentadecafluoro-1-octanol, 1h,1h-perfluoro-1-octanol, 1h,1h-perfluorooctanol, pentadecafluorooctyl alcohol, unii-9x5w7fq62r, 1-octanol, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecafluoro, pentadecafluoro-1-octanol, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecafluoro-1-octanol, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecafluorooctyl alcohol, perfluorooctanol | |
| C(C(C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)O | |
| 400.087 | |
| 400.09 | |
| Crystalline Powder |
| 45°C | |
| C8H3F15O | |
| 5 g | |
| PJDOLCGOTSNFJM-UHFFFAOYSA-N | |
| 2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecafluorooctan-1-ol | |
| 67543 | |
| ≥98.0% (GC) | |
| 1H,1H-Pentadecafluoro-1-octanol |
Safety and Handling
TSCA : Yes