missing translation for 'onlineSavingsMsg'
Learn More

1H,1H-Pentadecafluoro-1-octanol 98.0+%, TCI America™

Catalog No. P09045G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P09045G 5 g
P090425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P09045G Supplier TCI America Supplier No. P09045G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 307-30-2
Molecular Formula C8H3F15O
Molecular Weight (g/mol) 400.087
MDL Number MFCD00004675
InChI Key PJDOLCGOTSNFJM-UHFFFAOYSA-N
Synonym 1h,1h-pentadecafluoro-1-octanol, 1h,1h-perfluoro-1-octanol, 1h,1h-perfluorooctanol, pentadecafluorooctyl alcohol, unii-9x5w7fq62r, 1-octanol, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecafluoro, pentadecafluoro-1-octanol, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecafluoro-1-octanol, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecafluorooctyl alcohol, perfluorooctanol
PubChem CID 67543
IUPAC Name 2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecafluorooctan-1-ol
SMILES C(C(C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)O

Specifications

Melting Point 45°C
Boiling Point 164°C
Quantity 5 g
Formula Weight 400.09
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1H,1H-Pentadecafluoro-1-octanol