missing translation for 'onlineSavingsMsg'
Learn More

1H,1H-Nonafluoro-1-pentanol 97.0+%, TCI America™

Catalog No. p-7133982
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N08101G 1 g
N08105G 5 g
N081025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. N08101G Supplier TCI America Supplier No. N08101G
Only null left

Chemical Identifiers

CAS 355-28-2
Molecular Formula C5H3F9O
Molecular Weight (g/mol) 250.064
MDL Number MFCD00236714
InChI Key PJRIQFXPYMVWOU-UHFFFAOYSA-N
Synonym 1H,1H-Perfluoro-1-pentanol, (Perfluorobutyl)methanol
PubChem CID 2782394
IUPAC Name 2,2,3,3,4,4,5,5,5-nonafluoropentan-1-ol
SMILES C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)O

Specifications

Color Colorless
Quantity 1 g
Formula Weight 250.06
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 1H,1H-Nonafluoro-1-pentanol