missing translation for 'onlineSavingsMsg'
Learn More

1H,1H-Heptafluorobutylamine 95.0+%, TCI America™

Catalog No. H13001G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H13001G 1 g
H13005G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H13001G Supplier TCI America Supplier No. H13001G

Chemical Identifiers

CAS 374-99-2
Molecular Formula C4H4F7N
Molecular Weight (g/mol) 199.072
MDL Number MFCD00014817
InChI Key WBGBQSRNXPVFDB-UHFFFAOYSA-N
Synonym 2,2,3,3,4,4,4-heptafluorobutylamine, 1h,1h-heptafluorobutylamine, heptafluorobutylamine, butylamine, 2,2,3,3,4,4,4-heptafluoro, 1-butanamine, 2,2,3,3,4,4,4-heptafluoro, 1h,1h-perfluorobutylamine, acmc-1af94, 1-butanamine,2,2,3,3,4,4,4-heptafluoro
PubChem CID 67807
IUPAC Name 2,2,3,3,4,4,4-heptafluorobutan-1-amine
SMILES C(C(C(C(F)(F)F)(F)F)(F)F)N

Specifications

Color Yellow
Boiling Point 69°C
Quantity 1 g
UN Number 2735
Formula Weight 199.07
Percent Purity ≥95.0% (T)
Physical Form Liquid
Chemical Name or Material 1H,1H-Heptafluorobutylamine
RTECSNumber EO5190000
TSCA No