missing translation for 'onlineSavingsMsg'
Learn More

1H,1H,5H-Octafluoropentyl Acrylate (stabilized with MEHQ) 97.0+%, TCI America™

Catalog No. O03185G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O03185G 5 g
O031825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O03185G Supplier TCI America Supplier No. O03185G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 376-84-1
Molecular Formula C8H6F8O2
Molecular Weight (g/mol) 286.121
InChI Key WISUNKZXQSKYMR-UHFFFAOYSA-N
Synonym 1h,1h,5h-octafluoropentyl acrylate, 2,2,3,3,4,4,5,5-octafluoropentyl acrylate, 2-propenoic acid, 2,2,3,3,4,4,5,5-octafluoropentyl ester, acrylic acid 1h,1h,5h-octafluoropentyl ester, wisunkzxqskymr-uhfffaoysa, 2,2,3,3,4,4,5,5-octafluoropentylacrylate, acrylic acid=2,2,3,3,4,4,5,5-octafluoropentyl ester, 1h,1h,5h-octafluoropentyl acrylate stabilized with mehq, 2-propenoic acid,2,2,3,3,4,4,5,5-octafluoropentyl ester, 2,2,3,3,4,4,5,5-octafluoropentyl acrylate, contains 100 ppm monomethyl ether hydroquinone as inhibitor
PubChem CID 67829
IUPAC Name 2,2,3,3,4,4,5,5-octafluoropentyl prop-2-enoate
SMILES C=CC(=O)OCC(C(C(C(F)F)(F)F)(F)F)(F)F

Specifications

Color Colorless
Boiling Point 84°C
Quantity 5 g
Formula Weight 286.12
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 1H,1H,5H-Octafluoropentyl Acrylate (stabilized with MEHQ)
TSCA No
Recommended Storage Refrigerator