missing translation for 'onlineSavingsMsg'
Learn More

1H,1H,2H-Perfluoro-1-octene, 99%

Catalog No. AAB2092006
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2092006 5 g
AAB2092014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2092006 Supplier Thermo Scientific Chemicals Supplier No. B2092006
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 25291-17-2 | C8H3F13 | 346.091 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 25291-17-2
Molecular Formula C8H3F13
Molecular Weight (g/mol) 346.091
MDL Number MFCD00039249
InChI Key FYQFWFHDPNXORA-UHFFFAOYSA-N
Synonym perfluorohexyl ethylene, 1h,1h,2h-perfluoro-1-octene, tridecafluorohexyl ethylene, 1-octene, 3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluoro, unii-j4kbi9s48d, 3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluoro-1-octene, j4kbi9s48d, perfluorohexylethylene, acmc-209gix, c6f13ch=ch2
PubChem CID 91384
IUPAC Name 3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooct-1-ene
SMILES C=CC(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F

Specifications

Density 1.58
Boiling Point 106°C to 107°C
Flash Point 20°C (202°F)
Refractive Index 1.296
Quantity 5 g
Beilstein 2060994
Formula Weight 346.09
Percent Purity 99%
Chemical Name or Material 1H,1H,2H-Perfluoro-1-octene
Hazard Category H224-H315-H319-H335
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Hazard Class: 3; Packaging Group: II
EINECSNumber 246-791-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only