missing translation for 'onlineSavingsMsg'
Learn More

1H,1H,2H-Perfluoro-1-dodecene, 97%

Catalog No. AAL1659114
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1659114 25 g
AAL1659106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1659114 Supplier Thermo Scientific Chemicals Supplier No. L1659114
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 30389-25-4 | C12H3F21 | 546.122 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 30389-25-4
Molecular Formula C12H3F21
Molecular Weight (g/mol) 546.122
MDL Number MFCD00042346
InChI Key UCHSAVGOZUCXHC-UHFFFAOYSA-N
Synonym perfluorodecyl ethylene, 1h,1h,2h-perfluoro-1-dodecene, acmc-20aplc, n-perfluorodecyl ethylene, heneicosafluorodecyl-ethylene, 1h,1h,2h-perfluorododec-1-ene, heneicosafluorodecyl ethylene
PubChem CID 121692
IUPAC Name 3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,12-henicosafluorododec-1-ene
SMILES C=CC(C(C(C(C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F

Specifications

Density 1.711
Boiling Point 71°C to 72°C (14 mmHg)
Refractive Index 1.305
Linear Formula CF3(CF2)9CH=CH2
Quantity 25 g
Beilstein 2493721
Formula Weight 546.12
Percent Purity 97%
Chemical Name or Material 1H,1H,2H-Perfluoro-1-dodecene
EINECSNumber 250-173-3
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only