missing translation for 'onlineSavingsMsg'
Learn More

1H,1H,2H,2H-Tridecafluoro-n-octyl Methacrylate (stabilized with HQ + MEHQ) 98.0+%, TCI America™

Supplier:  TCI America T3414100G

 View more versions of this product

Catalog No. T3414100G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

1H,1H,2H,2H-Tridecafluoro-n-octyl Methacrylate (stabilized with HQ + MEHQ)
92°C
MFCD00077580
Methacrylic Acid 1H,1H,2H,2H-Tridecafluoro-n-octyl Ester, 3,3,4,4,5,5,6,6,7,7,8,8,8-Tridecafluoro-n-octyl Methacrylate, Methacrylic Acid 3,3,4,4,5,5,6,6,7,7,8,8,8-Tridecafluoro-n-octyl Ester
CC(=C)C(=O)OCCC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F
432.18
432.18
Liquid
2144-53-8
C12H9F13O2
100 g
CDXFIRXEAJABAZ-UHFFFAOYSA-N
3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl 2-methylprop-2-enoate
75066
≥98.0% (GC)

Chemical Identifiers

2144-53-8
432.18
CDXFIRXEAJABAZ-UHFFFAOYSA-N
75066
CC(=C)C(=O)OCCC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F
C12H9F13O2
MFCD00077580
Methacrylic Acid 1H,1H,2H,2H-Tridecafluoro-n-octyl Ester, 3,3,4,4,5,5,6,6,7,7,8,8,8-Tridecafluoro-n-octyl Methacrylate, Methacrylic Acid 3,3,4,4,5,5,6,6,7,7,8,8,8-Tridecafluoro-n-octyl Ester
3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl 2-methylprop-2-enoate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-3492&(2)-4065

RTECSNumber : UD3580000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions