missing translation for 'onlineSavingsMsg'
Learn More

1H,1H,2H,2H-Tridecafluoro-1-n-octanol 98.0+%, TCI America™

Catalog No. p-7137265
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T25285G 5 g
T252825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T25285G Supplier TCI America Supplier No. T25285G
Only null left

Chemical Identifiers

CAS 647-42-7
Molecular Formula C8H5F13O
Molecular Weight (g/mol) 364.106
MDL Number MFCD00042143
InChI Key GRJRKPMIRMSBNK-UHFFFAOYSA-N
Synonym 3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluoro-1-octanol, 2-perfluorohexyl ethanol, 1h,1h,2h,2h-perfluorooctan-1-ol, 1h,1h,2h,2h-perfluorooctanol, 1h,1h,2h,2h-perfluoro-1-octanol, unii-g2r5yo5n3v, 1-octanol, 3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluoro, 1h,1h,2h,2h-tridecafluoro-1-n-octanol, g2r5yo5n3v, 1,1,2,2-tetrahydroperfluoro-1-octanol
PubChem CID 69537
IUPAC Name 3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctan-1-ol
SMILES C(CO)C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F

Specifications

Color Yellow
Boiling Point 87°C
Quantity 5 g
Formula Weight 364.11
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1H,1H,2H,2H-Tridecafluoro-1-n-octanol
EINECSNumber (2)-2402
TSCA Yes