missing translation for 'onlineSavingsMsg'
Learn More

1H,1H,2H,2H-Perfluorooctyltriethoxysilane, 97%

Catalog No. AAB2462006
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2462006 5 g
AAB2462014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2462006 Supplier Thermo Scientific Chemicals Supplier No. B2462006
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 51851-37-7 | C14H19F13O3Si | 510.367 g/mol

1H,1H,2H,2H-Perfluorooctyltriethoxysilane is used as an active component of fluorine modified ORMOCER (organically modified ceramics) coating materials. It is a fluorine based polymer having low surface free energy, enhances its anti-adhesive behavior of polar and non-polar substances. Its wetting property acts in repelling aqueous electrolyte solution away from metallic substrates, thereby giving protection to metal substrate from corrosion. It is utilized in the surface modification of glass nanoparticles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1H,1H,2H,2H-Perfluorooctyltriethoxysilane is used as an active component of fluorine modified ORMOCER (organically modified ceramics) coating materials. It is a fluorine based polymer having low surface free energy, enhances its anti-adhesive behavior of polar and non-polar substances. Its wetting property acts in repelling aqueous electrolyte solution away from metallic substrates, thereby giving protection to metal substrate from corrosion. It is utilized in the surface modification of glass nanoparticles.

Solubility
Miscible with alcohol, aromatic and aliphatic hydrocarbons.

Notes
Moisture sensitive. Incompatible with moisture and oxidizing agents.

Chemical Identifiers

CAS 51851-37-7
Molecular Formula C14H19F13O3Si
Molecular Weight (g/mol) 510.367
MDL Number MFCD00042333
InChI Key AVYKQOAMZCAHRG-UHFFFAOYSA-N
Synonym 1h,1h,2h,2h-perfluorooctyltriethoxysilane, triethoxy 3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl silane, silane, triethoxy 3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl, perfluorooctyl triethoxysilane, 3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyltriethoxysilan, triethoxy 1h,1h,2h,2h-perfluoro-1-octyl silane, tridecafluoro-1,1,2,2-tetrahydrooctyl triethoxysilane, 1h,1h,2h,2h-perfluoroalkyltriethoxysilane, triethoxy-1h,1h,2h,2h-perfluoro-n-octylsilane
PubChem CID 103991
IUPAC Name triethoxy(3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl)silane
SMILES CCO[Si](CCC(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(OCC)OCC

Specifications

Boiling Point 220°C
Melting Point <-38°C
Quantity 5 g
Linear Formula CF3(CF2)5(CH2)2Si(OCH2CH3)3
Flash Point 97°C (206°F)
Solubility Information Miscible with alcohol,aromatic and aliphatic hydrocarbons.
Formula Weight 510.37
Percent Purity 97%
Refractive Index 1.345
Sensitivity Moisture sensitive
Density 1.333
Chemical Name or Material 1H,1H,2H,2H-Perfluorooctyltriethoxysilane
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
EINECSNumber 257-473-3
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only