missing translation for 'onlineSavingsMsg'
Learn More

1H,1H,2H,2H-Perfluorooctyltrichlorosilane, 97%

Catalog No. AAL1660618
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1660618 50 g
AAL1660609 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1660618 Supplier Thermo Scientific Chemicals Supplier No. L1660618
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 78560-45-9 | C8H4Cl3F13Si | 481.534 g/mol

1H,1H,2H,2H-Perfluorooctyltrichlorosilane is used in the silanization of silicon wafer. Self assembled monolayers treated with this, acts as an anti adhesive layer and release of cured poly(dimethyl siloxanes). It is also used to fabricate chemical surface patterns of self assembled monolayers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1H,1H,2H,2H-Perfluorooctyltrichlorosilane is used in the silanization of silicon wafer. Self assembled monolayers treated with this, acts as an anti adhesive layer and release of cured poly(dimethyl siloxanes). It is also used to fabricate chemical surface patterns of self assembled monolayers.

Solubility
Miscible with tetrahydrofuran, tetrhydropyran, toluene and other organic solvents.

Notes
Incompatible with water, strong oxidizing agents, alcohols, acids and bases.

Chemical Identifiers

CAS 78560-45-9
Molecular Formula C8H4Cl3F13Si
Molecular Weight (g/mol) 481.534
MDL Number MFCD00042363
InChI Key PISDRBMXQBSCIP-UHFFFAOYSA-N
Synonym trichloro 3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl silane, 1h,1h,2h,2h-perfluorooctyltrichlorosilane, silane, trichloro 3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl, tridecafluoro-1,1,2,2-tetrahydrooctyl trichlorosilane, trichloro 1h,1h,2h,2h-perfluorooctyl silane, 2-tridecafluorohexyl ethyltrichlorosilane, 1h,1h,2h,2h-perfluorooctyl trichlorosilane, trichloro 1h,1h,2h,2h-perfluoro-n-octyl silane, trichloro 1h,1h,2h,2h-tridecafluoro-n-octyl silane
PubChem CID 123578
IUPAC Name trichloro(3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl)silane
SMILES C(C[Si](Cl)(Cl)Cl)C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F

Specifications

Boiling Point 192°C
Quantity 50 g
UN Number UN2987
Beilstein 6154526
Flash Point 87°C (189°F)
Solubility Information Miscible with tetrahydrofuran,tetrhydropyran,toluene and other organic solvents.
Formula Weight 481.55
Percent Purity 97%
Refractive Index 1.352
Sensitivity Moisture sensitive
Density 1.638
Chemical Name or Material 1H,1H,2H,2H-Perfluorooctyltrichlorosilane
Show More Show Less
Hazard Category H227-H314-H335
Hazard Statement GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Precautionary Statement P210-P235-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CHLOROSILANES, CORROSIVE, N.O.S.
EINECSNumber 278-947-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only